Learn More
Mouse anti-Rat IgG1 Heavy Chain Secondary Antibody (MARG1-2), Janelia Fluor 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-68397JF646
Product Code. 30231546
  • £532.00 / 0.25mL
Description
IgG1 Heavy Chain Monoclonal antibody specifically detects IgG1 Heavy Chain in Rat samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG1 Heavy Chain
Monoclonal
Janelia Fluor 646
Mouse
Affinity purified
RUO
Rat
Purified
ELISA
MARG1-2
50mM Sodium Borate
Rat IR27 IgG1 myeloma protein
0.25 mL
Secondary
Store at 4°C in the dark.
IgG1