missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse anti-Rat Kappa Light Chain Secondary Antibody
Mouse Monoclonal Secondary Antibody
£280.00 - £337.00
Specifications
| Antigen | Rat Kappa Light Chain |
|---|---|
| Clone | OX-12 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
11545130
|
Invitrogen™
SA170012 |
250 μg |
£280.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15356321
|
Invitrogen™
SA510177 |
500 μg |
£337.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mouse anti-Rat Kappa Light Chain Secondary Antibody for Flow
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
| Rat Kappa Light Chain | |
| 1 mg/mL | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 0.02% sodium azide | |
| Pooled rat serum IgG. | |
| Secondary Antibody |
| OX-12 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG2a | |
| Rat | |
| Iglc | |
| Secondary | |
| Protein G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title