missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Rat IgE Heavy Chain Secondary Antibody (MARE-1), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-68394APC
This item is not returnable.
View return policy
Description
IgE Heavy Chain Monoclonal antibody specifically detects IgE Heavy Chain in Rat samples. It is validated for Western Blot, ELISA
Specifications
| IgE Heavy Chain | |
| Monoclonal | |
| APC | |
| Constant Region of Heavy Chain of IgE, Ig Epsilon Chain C Region, Ig Epsilon Chain C Region ND, IgE, Immunoglobulin Epsilon, immunoglobulin heavy constant epsilon | |
| Rat IR162, IR1016, IR2 and IR410 IgE myeloma proteins | |
| 0.25 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, ELISA | |
| MARE-1 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction