Learn More
RARRES2, Mouse, Clone: 3D8, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant RARRES2.
Brand: Abnova H00005919-M24.100ug
Description
This gene encodes a secreted chemotactic protein that initiates chemotaxis via the ChemR23 G protein-coupled seven-transmembrane domain ligand. Expression of this gene is upregulated by the synthetic retinoid tazarotene and occurs in a wide variety of tissues. The active protein has several roles, including that as an adipokine, and is truncated on both termini from the proprotein. [provided by RefSeq
Sequence: VGVAELTEAQRRGLQVALEEFHKHPPVQWAFQETSVESAVDTPFPAGIFVRLEFKLQQTSCRKRDWKKPECKVRPNGRKRKCLACIKLGSEDKVLGRLVHCPIETQVLREAEEHQETQCLRVQRAGEDPHSFYFPGQFAFSKALPRSSpecifications
| RARRES2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_002889.2 | |
| RARRES2 | |
| RARRES2 (NP_002880.1,17 a.a. ∽ 163 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA | |
| 3D8 | |
| Mouse monoclonal antibody raised against a full-length recombinant RARRES2. | |
| RARRES2 | |
| CHEMERIN/HP10433/TIG2 | |
| Mouse | |
| Protein A Purification | |
| RUO | |
| 5919 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.