Learn More
RARA, Mouse, Clone: 763, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of RARA.
Brand: Abnova MAB1779.100uL
Description
Retinoid signaling is transduced by 2 families of nuclear receptors, retinoic acid receptor (RAR) and retinoid X receptor (RXR; see MIM 180245), which form RXR/RAR heterodimers. In the absence of ligand, DNA-bound RXR/RARA represses transcription by recruiting the corepressors NCOR1 (MIM 600849), SMRT (NCOR2; MIM 600848), and histone deacetylase (see MIM 601241). When ligand binds to the complex, it induces a conformational change allowing the recruitment of coactivators, histone acetyltransferases (see MIM 603053), and the basic transcription machinery. Translocations that always involve rearrangement of the RARA gene are a cardinal feature of acute promyelocytic leukemia (APL; MIM 612376). The most frequent translocation is t(15,17)(q21;q22), which fuses the RARA gene with the PML gene (MIM 102578) (Vitoux et al., 2007 [PubMed 17468032]).[supplied by OMIM
Specifications
| RARA | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| RARA | |
| RARA | |
| A synthetic peptide corresponding to N-terminus human RARA. | |
| 100 μL | |
| Primary | |
| Specific for the ∼48k RARB isotype. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Western Blot | |
| 763 | |
| Mouse monoclonal antibody raised against synthetic peptide of RARA. | |
| In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 10% BSA) | |
| NR1B1/RAR | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 5914 | |
| Human | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.