Learn More
Mouse anti-Rabbit IgG (H+L) Secondary Antibody, FITC (Pre-adsorbed), Novus Biologicals™
Mouse Polyclonal Antibody
Brand:  Novus Biologicals NBP1-73657
Product Code. 18252052
  • £305.00 / 1mg
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG (H+L)
Polyclonal
Fluorescein
Mouse
Affinity Purified
RUO
Secondary
Reconstitute with 1.0 ml deionized water (or equivalent).
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
LYOPH
Western Blot 1:1000 - 1:10000, Flow Cytometry 1:500 - 1:2500, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000
Rabbit IgG whole molecule
1 mg
Adaptive Immunity, Immunology
This antibody was pre-adsorbed against Human, Goat, and Mouse Serum Proteins. No reaction was observed against Human, Goat and Mouse Serum Proteins.