Learn More
RAB25, Mouse, Clone: 3F12, Abnova™
Mouse monoclonal antibody raised against recombinant RAB25.
Brand: Abnova MAB10406.100uL
Description
RAB proteins, such as RAB25, are members of the RAS superfamily of small GTPases that are involved in membrane trafficking. Members of the RAB11 subfamily, including RAB25, control the return of internalized membrane-associated moieties to the cell surface (Caswell et al., 2007 [PubMed 17925226]).[supplied by OMIM
Specifications
| RAB25 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. | |
| RAB25 | |
| RAB25 | |
| Recombinant protein corresponding to human RAB25. | |
| RUO | |
| 57111 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 3F12 | |
| Mouse monoclonal antibody raised against recombinant RAB25. | |
| In ascites (0.03% sodium azide) | |
| CATX-8 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.