Learn More
PTGES2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant PTGES2.
Brand: Abnova H00080142-A01.50uL
Description
The protein encoded by this gene is a membrane-associated prostaglandin E synthase, which catalyzes the conversion of prostaglandin H2 to prostaglandin E2. This protein also has been shown to activate the transcription regulated by a gamma-interferon-activated transcription element (GATE). Multiple transcript variants have been found for this gene. [provided by RefSeq
Sequence: FGAVEGAVAKYMGAAAMYLISKRLKSRHRLQDNVREDLYEAADKWVAAVGKDRPFMGGQKPNLADLAVYGVLRVMEGLDAFDDLMQHTHIQPWYLRVERAITEASPAHSpecifications
| PTGES2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant PTGES2. | |
| PTGES2 | |
| C9orf15/FLJ14038/GBF1/MGC11289/PGES2/mPGES-2 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_025072 | |
| PTGES2 | |
| PTGES2 (NP_079348, 270 a.a. ∽ 377 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 80142 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only