Learn More
PSPH, Mouse, Clone: 3G12, Abnova™
Mouse monoclonal antibody raised against full-length recombinant PSPH.
Brand: Abnova MAB1093.100uL
Description
The protein encoded by this gene belongs to a subfamily of the phosphotransferases. This encoded enzyme is responsible for the third and last step in L-serine formation. It catalyzes magnesium-dependent hydrolysis of L-phosphoserine and is also involved in an exchange reaction between L-serine and L-phosphoserine. Deficiency of this protein is thought to be linked to Williams syndrome. [provided by RefSeq
Specifications
| PSPH | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user. | |
| PSPH | |
| PSP | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
| ELISA, Western Blot | |
| 3G12 | |
| Mouse monoclonal antibody raised against full length recombinant PSPH. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| NM_004577 | |
| PSPH | |
| Recombinant protein corresponding to full length human PSPH. | |
| RUO | |
| 5723 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.