missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSE1/KU, Mouse, Clone: N9C1, Abnova™
Mouse monoclonal antibody raised against native PSE1/KU.
Brand: Abnova MAB4320.200ug
This item is not returnable.
View return policy
Specifications
| PSE1/KU | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 200 μg | |
| Primary | |
| Human | |
| Liquid |
| Western Blot | |
| N9C1 | |
| Mouse monoclonal antibody raised against native PSE1/KU. | |
| In PBS, pH 7.4 | |
| Native purified PSE1/KU from human placenta extracted U1 small nuclear RNA. | |
| RUO | |
| This product recognizes the large alpha subunit (∼80-90 KDa) of placental PSE1 and human Ku in solution and when bound to DNA. | |
| Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction