missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRTN3, Mouse, Clone: 3B4, Abnova™
Mouse monoclonal antibody raised against recombinant PRTN3.
Brand: Abnova MAB10304.100uL
This item is not returnable.
View return policy
Specifications
| PRTN3 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| PRTN3 | |
| PRTN3 | |
| Recombinant protein corresponding to human PRTN3. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 3B4 | |
| Mouse monoclonal antibody raised against recombinant PRTN3. | |
| In PBS (50% glycerol, 0.03% sodium azide) | |
| ACPA/AGP7/C-ANCA/MBT/P29/PR-3 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 5657 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction