missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prolactin, Mouse, Clone: 6F11, Abnova™
Mouse monoclonal antibody raised against Prolactin.
Brand: Abnova MAB2284.100ug
This item is not returnable.
View return policy
Specifications
| Prolactin | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| PRL | |
| A mixture native PRL of sheep and human. | |
| RUO | |
| 5617/443317 | |
| Bovine, Human, Monkey, Mouse, Rat, Sheep | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 6F11 | |
| Mouse monoclonal antibody raised against Prolactin. | |
| In PBS, pH 7.4 | |
| Mouse | |
| 100 μg | |
| Primary | |
| The epitope recognized by this antibody is restricted to a region that includes the carboxyl-terminal disulfide loop that is conserved among prolactins from several species. | |
| Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction