Learn More
PPP1CA Mouse anti-Human, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against partial recombinant protein of PPP1CA.
Brand: Abnova MAB11159.50ug
Description
The protein encoded by this gene is one of the three catalytic subunits of protein phosphatase 1 (PP1). PP1 is a serine/threonine specific protein phosphatase known to be involved in the regulation of a variety of cellular processes, such as cell division, glycogen metabolism, muscle contractility, protein synthesis, and HIV-1 viral transcription. Increased PP1 activity has been observed in the end stage of heart failure. Studies in both human and mice suggest that PP1 is an important regulator of cardiac function. Mouse studies also suggest that PP1 functions as a suppressor of learning and memory. Three alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| PPP1CA | |
| Monoclonal | |
| Mouse monoclonal antibody raised against partial recombinant protein of PPP1CA. | |
| In PBS, pH 7.4 (1% BSA, 50% glycerol, 0.02% sodium azide) | |
| MGC15877/MGC1674/PP-1A/PPP1A | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 5499 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:5000-1:20000) Western Blot (1:100-1:2000) The optimal working dilution should be determined by the end user. | |
| PPP1CA | |
| PPP1CA | |
| Recombinant protein corresponding to N-terminus of human PPP1CA. | |
| 50 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.