Learn More
PPIB, Mouse, Clone: k2E2, Abnova™
Mouse monoclonal antibody raised against partial recombinant PPIB.
Brand: Abnova MAB2033.100uL
Description
The protein encoded by this gene is a cyclosporine-binding protein and is mainly located within the endoplasmic reticulum. It is associated with the secretory pathway and released in biological fluids. This protein can bind to cells derived from T- and B-lymphocytes, and may regulate cyclosporine A-mediated immunosuppression. [provided by RefSeq
Specifications
| PPIB | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| PPIB | |
| PPIB | |
| Recombinant protein corresponding to amino acids 26-216 of human PPIB. | |
| RUO | |
| 5479 | |
| Liquid |
| ELISA, Western Blot | |
| k2E2 | |
| Mouse monoclonal antibody raised against partial recombinant PPIB. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| CYP-S1/CYPB/MGC14109/MGC2224/SCYLP | |
| Mouse | |
| 100 μL | |
| Primary | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.