Learn More
POP4, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant POP4.
Brand: Abnova H00010775-A01.50uL
Description
This gene encodes one of the protein subunits of the small nucleolar ribonucleoprotein complexes: the endoribonuclease for mitochondrial RNA processing complex and the ribonuclease P complex. The encoded protein is localized to the nucleus and associates directly with the RNA component of these complexes. This protein is involved in processing of precursor RNAs. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Sequence: TQPQMIQAKLLKADLHGAIISVTKSKCPSYVGITGILLQETKHIFKIITKEDRLKVIPKLNCVFTVETDGFISYIYGSKFQLRSSERSAKKFKAKGTIDLSpecifications
| POP4 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant POP4. | |
| POP4 | |
| RPP29 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_006627 | |
| POP4 | |
| POP4 (NP_006618, 121 a.a. ∽ 220 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 10775 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only