Learn More
PNLIP Mouse anti-Human, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against native PNLIP.
Brand: Abnova MAB2206.100ug
Description
This gene is a member of the lipase gene family. It encodes a carboxyl esterase that hydrolyzes insoluble, emulsified triglycerides, and is essential for the efficient digestion of dietary fats. This gene is expressed specifically in the pancreas. [provided by RefSeq
Specifications
| PNLIP | |
| Monoclonal | |
| Mouse monoclonal antibody raised against native PNLIP. | |
| In PBS, pH 7.4 | |
| PL | |
| Mouse | |
| 100 μg | |
| Primary | |
| These antibodies specifically recognize pancreatic lipase. | |
| Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Radioimmune Assays (RIA) | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| PNLIP | |
| PNLIP | |
| Native purified PNLIP from human pancreas. | |
| RUO | |
| 5406 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.