missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLXDC1, Mouse, Clone: 197C193 (IM193), Abnova™
Mouse monoclonal antibody raised against synthetic peptide of PLXDC1.
Brand: Abnova MAB0098.100ug
This item is not returnable.
View return policy
Specifications
| PLXDC1 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| PLXDC1 | |
| PLXDC1 | |
| A synthetic peptide corresponding to amino acids 411-427 of human PLXDC1. | |
| RUO | |
| 57125 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot | |
| 197C193 (IM193) | |
| Mouse monoclonal antibody raised against synthetic peptide of PLXDC1. | |
| In PBS (0.05% BSA, 0.05% sodium azide) | |
| DKFZp686F0937/FLJ36270/FLJ45632/TEM3/TEM7 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction