missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plat, Mouse, Clone: H27B6, Abnova™
Mouse monoclonal antibody raised against recombinant Plat.
Brand: Abnova MAB3790.1mg
This item is not returnable.
View return policy
Specifications
| Plat | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| Plat | |
| Plat | |
| Recombinant protein corresponding to mouse Plat. | |
| RUO | |
| 18791 | |
| Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunohistochemistry, Western Blot | |
| H27B6 | |
| Mouse monoclonal antibody raised against recombinant Plat. | |
| In 50mM sodium phosphate buffer, 150mM NaCl, pH 7.4 | |
| AU020998/AW212668/D8Ertd2e/MGC18508/t-PA/tPA | |
| Mouse | |
| 1 mg | |
| Primary | |
| Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction