Learn More
PEX16, Mouse, Clone: 3E10, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant PEX16.
Brand: Abnova H00009409-M01A.200uL
Description
The protein encoded by this gene is an integral peroxisomal membrane protein. An inactivating nonsense mutation localized to this gene was observed in a patient with Zellweger syndrome of the complementation group CGD/CG9. Expression of this gene product morphologically and biochemically restores the formation of new peroxisomes, suggesting a role in peroxisome organization and biogenesis. Alternative splicing has been observed for this gene and two variants have been described. [provided by RefSeq
Sequence: MEKLRLLGLRYQEYVTRHPAATAQLETAVRGFSYLLAGRFADSHELSELVYSASNLLVLLNDGILRKELRKKLPVSLSQQKLLTWLSVLECVEVFMEMGAAKVWGELGRWLVIALIQLAKAVLRILLLLWFKAGLQTSPPIAPLDRETQAQPPDGDHSPGNHEQSYVGKRSNRVVRTLQNTPSLHSRHWGAPQQREGRQQQHHEELSATPTPLGLQETIAEFLYIARPLLHLLSLGLWGQRSWKPWLLAGVVDVTSLSLLSDRKGLTRRERRELRRRTILLLYYLLRSPFYDRFSEARILFLLQLLADHVPGVGLVTTSQRAASPCLPARPHTQPWSPPAFLPGHPSpecifications
| PEX16 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC000467 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgM κ |
| ELISA | |
| 3E10 | |
| Mouse monoclonal antibody raised against a full-length recombinant PEX16. | |
| PEX16 | |
| PEX16 | |
| PEX16 (AAH00467,1 a.a. ∽ 346 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 9409 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.