Learn More
PEG10, Mouse, Clone: 4C10A7, Abnova™
Mouse monoclonal antibody raised against recombinant PEG10.
Brand: Abnova MAB10294.100uL
Description
This gene includes two overlapping reading frames of the same transcript encoding distinct isoforms. The shorter isoform has a CCHC-type zinc finger motif containing a sequence characteristic of gag proteins of most retroviruses and some retrotransposons, and it functions in part by interacting with members of the TGF-beta receptor family. The longer isoform has the active-site DSG consensus sequence of the protease domain of pol proteins. The longer isoform is the result of -1 translational frameshifting that is also seen in some retroviruses. Expression of these two isoforms only comes from the paternal allele due to imprinting. Increased gene expression (as observed by an increase in mRNA levels) is associated with hepatocellular carcinomas. [provided by RefSeq
Specifications
| PEG10 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
| PEG10 | |
| PEG10 | |
| Recombinant protein corresponding to human PEG10. | |
| RUO | |
| 23089 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| 4C10A7 | |
| Mouse monoclonal antibody raised against recombinant PEG10. | |
| In ascites (0.03% sodium azide) | |
| Edr/HB-1/KIAA1051/MEF3L/Mar2/Mart2/RGAG3 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.