missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PECAM1, Mouse, Clone: MEM-05, Abnova™
Mouse monoclonal antibody raised against native PECAM1.
Brand: Abnova MAB0913.100ug
This item is not returnable.
View return policy
Specifications
| PECAM1 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| PECAM1 | |
| PECAM1 | |
| Native purified PECAM1 from leukocytes of a patient suffering from a LGL-type leukemia. | |
| RUO | |
| 5175 | |
| Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Flow Cytometry, Western Blot | |
| MEM-05 | |
| Mouse monoclonal antibody raised against native PECAM1. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| CD31/PECAM-1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction