Learn More
PDGFRL, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant PDGFRL.
Brand: Abnova H00005157-A01.50uL
Description
This gene encodes a protein with significant sequence similarity to the ligand binding domain of platelet-derived growth factor receptor beta. Mutations in this gene, or deletion of a chromosomal segment containing this gene, are associated with sporadic hepatocellular carcinomas, colorectal cancers, and nonsmall cell lung cancers. This suggests this gene product may function as a tumor suppressor. [provided by RefSeq
Sequence: ILASSNKVKSGDDISVLCTVLGEPDVEVEFTWIFPGQKDERPVTIQDTWRLIHRGLGHTTRISQSVITVEDFETIDAGYYICTAQNLQGQTTVATTVEFSSpecifications
| PDGFRL | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant PDGFRL. | |
| PDGFRL | |
| PDGRL/PRLTS | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_006207 | |
| PDGFRL | |
| PDGFRL (NP_006198, 276 a.a. ∽ 375 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 5157 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only