Learn More
PDCD4, Mouse, Clone: 9G6, Abnova™
Mouse monoclonal antibody raised against synthetic phosphopeptide of PDCD4.
Brand: Abnova MAB10262.100ug
Description
This gene encodes a protein localized to the nucleus in proliferating cells. Expression of this gene is modulated by cytokines in natural killer and T cells. The gene product is thought to play a role in apoptosis but the specific role has not yet been determined. Two transcripts encoding different isoforms have been identified. [provided by RefSeq
Specifications
| PDCD4 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:20000-1:100000) Western Blot (1:2000-1:10000) Immunohistochemistry (1:1000-1:5000) The optimal working dilution should be determined by the end user. | |
| PDCD4 | |
| PDCD4 | |
| Synthetic phosphopeptide corresponding to residues surrounding S457 of human PDCD4. | |
| 100 μg | |
| Primary | |
| This antibody is specific for human Pdcd4 protein phosphorylated at Ser457. A BLAST analysis was used to suggest cross-reactivity with Pdcd4 from human, mouse, rat and Xenopus based on 100% homology with the immunizing sequence. Cross-reactivity with Pdcd4 from other sources has not been determined. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 9G6 | |
| Mouse monoclonal antibody raised against synthetic phosphopeptide of PDCD4. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| H731/MGC33046/MGC33047 | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 27250 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.