Learn More
PCNA, Mouse, Clone: k1A3, Abnova™
Mouse monoclonal antibody raised against full-length recombinant PCNA.
Brand: Abnova MAB3206.100uL
Description
The protein encoded by this gene is found in the nucleus and is a cofactor of DNA polymerase delta. The encoded protein acts as a homotrimer and helps increase the processivity of leading strand synthesis during DNA replication. In response to DNA damage, this protein is ubiquitinated and is involved in the RAD6-dependent DNA repair pathway. Two transcript variants encoding the same protein have been found for this gene. Pseudogenes of this gene have been described on chromosome 4 and on the X chromosome. [provided by RefSeq
Specifications
| PCNA | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| PCNA | |
| PCNA | |
| Recombinant protein corresponding to full length human PCNA. | |
| RUO | |
| 5111 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG3 κ |
| ELISA, Western Blot | |
| k1A3 | |
| Mouse monoclonal antibody raised against full length recombinant PCNA. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| MGC8367 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.