Learn More
PARP1, Mouse, Clone: 7A10, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of PARP1.
Brand: Abnova MAB10291.100uL
Description
This gene encodes a chromatin-associated enzyme, poly(ADP-ribosyl)transferase, which modifies various nuclear proteins by poly(ADP-ribosyl)ation. The modification is dependent on DNA and is involved in the regulation of various important cellular processes such as differentiation, proliferation, and tumor transformation and also in the regulation of the molecular events involved in the recovery of cell from DNA damage. In addition, this enzyme may be the site of mutation in Fanconi anemia, and may participate in the pathophysiology of type I diabetes. [provided by RefSeq
Specifications
| PARP1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. | |
| PARP1 | |
| PARP1 | |
| A synthetic peptide (conjugated with KLH) corresponding to human PARP1. | |
| RUO | |
| 142 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 7A10 | |
| Mouse monoclonal antibody raised against synthetic peptide of PARP1. | |
| In ascites (0.03% sodium azide) | |
| ADPRT/ADPRT1/PARP/PARP-1/PPOL/pADPRT-1 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.