Learn More
PAPPA, Mouse, Clone: 7A6, Abnova™
Mouse monoclonal antibody raised against native PAPPA.
Brand: Abnova MAB0588.200ug
Description
This gene encodes a secreted metalloproteinase which cleaves insulin-like growth factor binding proteins (IGFBPs). It is thought to be involved in local proliferative processes such as wound healing and bone remodeling. Low plasma level of this protein has been suggested as a biochemical marker for pregnancies with aneuploid fetuses. [provided by RefSeq
Specifications
| PAPPA | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| PAPPA | |
| PAPPA | |
| Native purified human PAPPA. | |
| RUO | |
| 5069 | |
| Store at 4°C. | |
| IgG2a |
| ELISA, Immunoprecipitation | |
| 7A6 | |
| Mouse monoclonal antibody raised against native PAPPA. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| ASBABP2/DIPLA1/IGFBP-4ase/PAPA/PAPP-A/PAPPA1 | |
| Mouse | |
| 200 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.