missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pan Cytokeratin, Mouse, Clone: AE1 + AE3, Abnova™
Mouse monoclonal antibody raised against Pan Cytokeratin.
Brand: Abnova MAB11311.100ug
This item is not returnable.
View return policy
Specifications
| Pan Cytokeratin | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (0.1 - 1.0 ug/ml) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200 - 1:400) The optimal working dilution should be determined by the end user. | |
| CK1/EHK1/K1/KRT1A | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 3848/3856/3880/51350 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
| Immunohistochemistry (PFA fixed), Western Blot | |
| AE1 + AE3 | |
| Mouse monoclonal antibody raised against Pan Cytokeratin. | |
| In PBS (0.05% BSA, 0.05% sodium azide) | |
| KRT1 | |
| 1982)., Total keratin isolated from human epidermal callus (Woodcock-Mitchell | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction