Learn More
PAFAH1B2, Mouse, Clone: 2F4-1C10, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant PAFAH1B2.
Brand: Abnova H00005049-M01A.200uL
Description
Platelet-activating factor (PAF) is a biologically active phospholipid with diverse biologic effects. PAF is degraded to inactive products by hydrolysis of the acetyl group at the sn-2 position to produce the biologically inactive products LYSO-PAF and acetate. This reaction is catalyzed by PAF acetylhydrolase (PAFAH). The various monomeric and multimeric forms of the enzyme are composed of alpha (MIM 601545), beta, and gamma (MIM 603074) PAFAH subunits.[supplied by OMIM
Sequence: MSQGDSNPAAIPHAAEDIQGDDRWMSQHNRFVLDCKDKEPDVLFVGDSMVQLMQQYEIWRELFSPLHALNFGIGGDTTRHVLWRLKNGELENIKPKVIVVWVGTNNHENTAEEVAGGIEAIVQLINTRQPQAKIIVLGLLPRGEKPNPLRQKNAKVNQLLKVSLPKLANVQLLDTDGGFVHSDGAISCHDMFDFLHLTGGGYAKICKPLHELIMQLLEETPEEKQTTIASpecifications
| PAFAH1B2 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC000398 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgM κ |
| ELISA, Western Blot | |
| 2F4-1C10 | |
| Mouse monoclonal antibody raised against a full-length recombinant PAFAH1B2. | |
| PAFAH1B2 | |
| PAFAH1B2 | |
| PAFAH1B2 (AAH00398,1 a.a. ∽ 229 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 5049 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.