Learn More
PABPC1, Mouse, Clone: 10E10, Abnova™
Mouse monoclonal antibody raised against recombinant PABPC1.
Brand: Abnova MAB2465.100uL
Description
The poly(A)-binding protein (PABP), which is found complexed to the 3-prime poly(A) tail of eukaryotic mRNA, is required for poly(A) shortening and translation initiation. In humans, the PABPs comprise a small nuclear isoform and a conserved gene family that displays at least 3 functional proteins: PABP1 (PABPC1), inducible PABP (iPABP, or PABPC4; MIM 603407), and PABP3 (PABPC3; MIM 604680). In addition, there are at least 4 pseudogenes, PABPCP1 to PABPCP4.[supplied by OMIM
Specifications
| PABPC1 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| PABPC1 | |
| PABPC1 | |
| Recombinant protein corresponding to human PABPC1. | |
| RUO | |
| 26986 | |
| Chicken, Frog, Human, Rabbit | |
| Liquid |
| ELISA, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 10E10 | |
| Mouse monoclonal antibody raised against recombinant PABPC1. | |
| In PBS (0.09% sodium azide) | |
| PAB1/PABP/PABP1/PABPC2/PABPL1 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Detects a band of approximately 70 KDa. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.