Learn More
OR13D1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant OR13D1.
Brand: Abnova H00286365-A01.50uL
Description
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq
Sequence: YRFTDFDVSNISIYLNHVLFYTTQQAGDLEHMETRNYSAMTEFFLVGLSQYPELQSpecifications
| OR13D1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant OR13D1. | |
| OR13D1 | |
| OR9-15 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_001004484 | |
| OR13D1 | |
| OR13D1 (NP_001004484, 2 a.a. ∽ 56 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 286365 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only