missing translation for 'onlineSavingsMsg'
Learn More

NSP1, Mouse, Clone: 32D6, Abnova™

Mouse monoclonal antibody raised against native NSP1.

Brand:  Abnova MAB3339.200uL

Product Code. 16065766

  • £643.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NSP1
Monoclonal
Unconjugated
Western Blot (1:500-1:10000) Immunocytochemistry (1:500-1:5000) Immunofluorescence (1:500-1:5000) The optimal working dilution should be determined by the end user.
NSP1
Mouse
200 μL
Primary
Yeast
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
32D6
Mouse monoclonal antibody raised against native NSP1.
In PBS (50% glycerol, 0.09% sodium azide)
NSP1
Native purified NSP1 from yeast nuclear protein preparations.
RUO
853409
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.