missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NQO2, Mouse, Clone: AT1E3, Abnova™
Mouse monoclonal antibody raised against partial recombinant NQO2.
Brand: Abnova MAB9720.100uL
This item is not returnable.
View return policy
Specifications
| NQO2 | |
| Monoclonal | |
| 1 mg/mL | |
| Mouse monoclonal antibody raised against partial recombinant NQO2. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| DHQV/DIA6/NMOR2/QR2 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 4835 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
| ELISA, Western Blot | |
| AT7G7 | |
| Unconjugated | |
| ELISA (1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| NQO2 | |
| NQO2 | |
| Recombinant protein corresponding to amino acids 1-231 of human NQO2. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction