missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ngfr, Mouse, Clone: MC192, Abnova™
Mouse monoclonal antibody raised against Ngfr.
Brand: Abnova MAB5595.100ug
This item is not returnable.
View return policy
Specifications
| Ngfr | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| Ngfr | |
| Ngfr | |
| Rat Ngfr. | |
| RUO | |
| 24596 | |
| Rat | |
| Lyophilized |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| MC192 | |
| Mouse monoclonal antibody raised against Ngfr. | |
| Lyophilized from cell culture supernatant (1% BSA) | |
| LNGFR/RNNGFRR/p75/p75NTR | |
| Mouse | |
| 100 μg | |
| Primary | |
| Specificity has been confirmed by immunohistochemical staining of lesioned sciatic nerve and spinal cord, the results reflect the reported literatures. | |
| Store at 4°C on dry atmosphere. After reconstitution with 100μL sterilzed water store at -20°C or lower. For an additional stability glycerol (1:1) may be added. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction