Learn More
NFKBIE, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant NFKBIE.
Brand: Abnova H00004794-A01.50uL
Description
NFKB1 (MIM 164011) or NFKB2 (MIM 164012) is bound to REL (MIM 164910), RELA (MIM 164014), or RELB (MIM 604758) to form the NFKB complex. The NFKB complex is inhibited by I-kappa-B proteins (NFKBIA, MIM 164008 or NFKBIB, MIM 604495), which inactivate NF-kappa-B by trapping it in the cytoplasm. Phosphorylation of serine residues on the I-kappa-B proteins by kinases (IKBKA, MIM 600664, or IKBKB, MIM 603258) marks them for destruction via the ubiquitination pathway, thereby allowing activation of the NF-kappa-B complex. Activated NFKB complex translocates into the nucleus and binds DNA at kappa-B-binding motifs such as 5-prime GGGRNNYYCC 3-prime or 5-prime HGGARNYYCC 3-prime (where H is A, C, or T; R is an A or G purine; and Y is a C or T pyrimidine). For some genes, activation requires NFKB interaction with other transcription factors, such as STAT (see STAT6, MIM 601512), AP1 (JUN, MIM 165160), and NFAT (see NFATC1, MIM 600489).[supplied by OMIM
Sequence: SGKTALHLAVETQERGLVQFLLQAGAQVDARMLNGCTPLHLAAGRGLMGISSTLCKAGADSLLRNVEDETPQDLTEESLVLLPFDDLKISGKLLLCSpecifications
| NFKBIE | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant NFKBIE. | |
| NFKBIE | |
| IKBE | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_004556 | |
| NFKBIE | |
| NFKBIE (NP_004547, 403 a.a. ∽ 498 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 4794 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only