Learn More
NEFH, Mouse, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against native NEFH.
Brand: Abnova MAB6235.100uL
Description
Neurofilaments are type IV intermediate filament heteropolymers composed of light, medium, and heavy chains. Neurofilaments comprise the axoskeleton and functionally maintain neuronal caliber. They may also play a role in intracellular transport to axons and dendrites. This gene encodes the heavy neurofilament protein. This protein is commonly used as a biomarker of neuronal damage and susceptibility to amyotrophic lateral sclerosis (ALS) has been associated with mutations in this gene. [provided by RefSeq
Specifications
| NEFH | |
| Monoclonal | |
| Mouse monoclonal antibody raised against native NEFH. | |
| In ascites | |
| NFH | |
| Mouse | |
| 100 μL | |
| Primary | |
| Bovine | |
| Liquid |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| Western Blot (1:5000) Immunofluorescence (1:500) The optimal working dilution should be determined by the end user. | |
| NEFH | |
| NEFH | |
| Native purified bovine NEFH. | |
| RUO | |
| 4744 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.