missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCOR2, Mouse, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of NCOR2.
Brand: Abnova MAB8270.100ug
This item is not returnable.
View return policy
Specifications
| NCOR2 | |
| Monoclonal | |
| Mouse monoclonal antibody raised against synthetic peptide of NCOR2. | |
| In PBS | |
| CTG26/SMRT/SMRTE/SMRTE-tau/TNRC14/TRAC-1/TRAC1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (1-5 ug/mL) Immunoprecipitation (1-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| NCOR2 | |
| NCOR2 | |
| A synthetic peptide corresponding to amino acids 994-1005 of human NCOR2. | |
| RUO | |
| 9612 | |
| Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction