missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCAPD2 (CNAP1), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CNAP1.
Brand: Abnova H00009918-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: LRKMLDNFDCFGDKLSDESIFSAFLSVVGKLRRGAKPEGKAIIDEFEQKLRACHTRGLDGIKELEIGQAGSQRAPSAKKPSSGSRYQPLASTASDNDFVTSpecifications
| NCAPD2 (CNAP1) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant CNAP1. | |
| NCAPD2 | |
| CAP-D2/CNAP1/KIAA0159/hCAP-D2 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC028182 | |
| NCAPD2 | |
| CNAP1 (AAH28182, 1240 a.a. ∽ 1339 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 9918 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto