missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCAM1, Mouse, Clone: 123A8, Abnova™
Mouse monoclonal antibody raised against human NCAM1.
Brand: Abnova MAB12140.100ug
This item is not returnable.
View return policy
Specifications
| NCAM1 | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (0.5-1 ug/mL) Western blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| NCAM1 | |
| NCAM1 | |
| Membrane preparation of human lung carcinoma cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 123A8 | |
| Mouse monoclonal antibody raised against human NCAM1. | |
| In PBS (0.05% BSA, 0.05% sodium azide) | |
| CD56/MSK39/NCAM | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 4684 | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction