missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCAM1, Mouse, Biotin, Clone: MEM-188, Abnova™
Mouse monoclonal antibody raised against native NCAM1.
Brand: Abnova MAB5002.100ug
This item is not returnable.
View return policy
Specifications
| NCAM1 | |
| Monoclonal | |
| Biotin | |
| Flow Cytometry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| NCAM1 | |
| NCAM1 | |
| Native purified NCAM1 from KG-1 human acute myelogenous leukemia cell line. | |
| RUO | |
| 4684 | |
| Human, Primate | |
| Liquid |
| Flow Cytometry | |
| MEM-188 | |
| Mouse monoclonal antibody raised against native NCAM1. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| CD56/MSK39/NCAM | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody reacts with a 180 KDa isoform of CD56 (NCAM) expressed in leukocytes. It has been suggested that this antibody could react with rhesus monkey lymphocytes. Reactivity with other NCAM isoforms has not been tested. | |
| Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction