missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Natural Killer Cell Activation Structures, Mouse, Clone: ANK61, Abnova™
Mouse monoclonal antibody raised against Natural Killer Cell Activation Structures.
Brand: Abnova MAB5198.1mL
This item is not returnable.
View return policy
Specifications
| Natural Killer Cell Activation Structures | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 1 mL | |
| Primary | |
| Rat | |
| Liquid |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation | |
| ANK61 | |
| Mouse monoclonal antibody raised against Natural Killer Cell Activation Structures. | |
| In culture supernatant (0.05% sodium azide) | |
| Native purified from IL-2-activated cultured NK cells of Wag rats. | |
| RUO | |
| ANK61 reactivity is rat strain-independent. The antigen recognized by ANK61 is highly expressed on freshly isolated and cultured rat NK cells. The antibodies also bind to rat alphabeta-TCR T cells and at a low level to rat B cells. Binding to other cell types is unknown. Triggering of the antigen by ANK61 antibodies activates the lytic machinery of rat NK cells, but not of T cells. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction