Learn More
NAMPT, Mouse, Clone: 4D5, Abnova™
Mouse monoclonal antibody raised against full-length recombinant NAMPT.
Brand: Abnova MAB1122.100uL
Description
This gene encodes a protein that catalyzes the condensation of nicotinamide with 5-phosphoribosyl-1-pyrophosphate to yield nicotinamide mononucleotide, one step in the biosynthesis of nicotinamide adenine dinucleotide. The protein is an adipokine that is localized to the bloodstream and has various functions, including the promotion of vascular smooth muscle cell maturation and inhibition of neutrophil apoptosis. It also activates insulin receptor and has insulin-mimetic effects, lowering blood glucose and improving insulin sensitivity. The protein is highly expressed in visceral fat and serum levels of the protein correlate with obesity. This gene has a pseudogene on chromosome 10. [provided by RefSeq
Specifications
| NAMPT | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:1000-1:3000) The optimal working dilution should be determined by the end user. | |
| NAMPT | |
| 1110035O14Rik/DKFZp666B131/MGC117256/PBEF/PBEF1/VF/VISFATIN | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 4D5 | |
| Mouse monoclonal antibody raised against full length recombinant NAMPT. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| NM_005746 | |
| NAMPT | |
| Recombinant protein corresponding to full length human NAMPT. | |
| RUO | |
| 10135 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.