missing translation for 'onlineSavingsMsg'
Learn More

MYO7A, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant MYO7A.

Brand:  Abnova H00004647-A01.50uL

Product Code. 16101615

  • £209.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene is a member of the myosin gene family. Myosins are mechanochemical proteins characterized by the presence of a motor domain, an actin-binding domain, a neck domain that interacts with other proteins, and a tail domain that serves as an anchor. This gene encodes an unconventional myosin with a very short tail. Defects in this gene are associated with the mouse shaker-1 phenotype and the human Usher syndrome 1B which are characterized by deafness, reduced vestibular function, and (in human) retinal degeneration. Alternative splicing results in multiple transcript variants. [provided by RefSeq

Sequence: KQTTEPNFPEILLIAINKYGVSLIDPKTKDILTTHPFTKISNWSSGNTYFHITIGNLVRGSKLLCETSLGYKMDDLLTSYISQMLTAMSKQRGSRS
Specifications

Specifications

MYO7A
Polyclonal
Mouse polyclonal antibody raised against a partial recombinant MYO7A.
MYO7A
DFNA11/DFNB2/MYOVIIA/MYU7A/NSRD2/USH1B
Mouse
50 μL
Primary
Human
Antisera
ELISA, Western Blot
Unconjugated
50% glycerol
NM_000260
MYO7A
MYO7A (NP_000251, 2118 a.a. ∽ 2213 a.a) partial recombinant protein with GST tag.
RUO
4647
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado

For Research Use Only