Learn More
MYL3, Mouse, Clone: 3F8, Abnova™
Mouse monoclonal antibody raised against recombinant MYL3.
Brand: Abnova MAB10429.100uL
Description
MYL3 encodes myosin light chain 3, an alkali light chain also referred to in the literature as both the ventricular isoform and the slow skeletal muscle isoform. Mutations in MYL3 have been identified as a cause of mid-left ventricular chamber type hypertrophic cardiomyopathy. [provided by RefSeq
Specifications
| MYL3 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user. | |
| MYL3 | |
| MYL3 | |
| Recombinant protein corresponding to human MYL3. | |
| RUO | |
| 4634 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| 3F8 | |
| Mouse monoclonal antibody raised against recombinant MYL3. | |
| In ascitic (0.03% sodium azide) | |
| CMH8/MLC1SB/MLC1V/VLC1 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.