Learn More
MYL2, Mouse, Clone: AT3B2, Abnova™
Mouse monoclonal antibody raised against full-length recombinant MYL2.
Brand: Abnova MAB5641.100uL
Description
Thus gene encodes the regulatory light chain associated with cardiac myosin beta (or slow) heavy chain. Ca+ triggers the phosphorylation of regulatory light chain that in turn triggers contraction. Mutations in this gene are associated with mid-left ventricular chamber type hypertrophic cardiomyopathy. [provided by RefSeq]
Specifications
| MYL2 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| MYL2 | |
| MYL2 | |
| Recombinant protein corresponding to full length human MYL2. | |
| RUO | |
| 4633 | |
| Store at 4°C. For long term storage store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b κ |
| ELISA, Western Blot | |
| AT3B2 | |
| Mouse monoclonal antibody raised against full length recombinant MYL2. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| CMH10/DKFZp779C0562/MLC2 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.