Learn More
MYL12B, Mouse, Clone: 401s3, Abnova™
Mouse monoclonal antibody raised against full-length recombinant MYL12B.
Brand: Abnova MAB8855.100uL
Description
The activity of nonmuscle myosin II (see MYH9; MIM 160775) is regulated by phosphorylation of a regulatory light chain, such as MRLC2. This phosphorylation results in higher MgATPase activity and the assembly of myosin II filaments (Iwasaki et al., 2001 [PubMed 11942626]).[supplied by OMIM
Specifications
| MYL12B | |
| Monoclonal | |
| 1 ug/uL | |
| Mouse monoclonal antibody raised against full length recombinant MYL12B. | |
| In citrate-Tris-HCl, pH7.0 (0.02% Proclin 300) | |
| MLC-B | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 103910 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 401s3 | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| MRLC2 | |
| MRLC2 | |
| Recombinant protein corresponding to full length of human MYL12B. | |
| 100 μL | |
| Primary | |
| This antibody recognizes recombinant protein MRLC2. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.