Learn More
MUC7, Mouse, Clone: 7F2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant MUC7.
Brand: Abnova H00004589-M06A.200uL
Description
This gene encodes a small salivary mucin, which is thought to play a role in facilitating the clearance of bacteria in the oral cavity and to aid in mastication, speech, and swallowing. The central domain of this glycoprotein contains tandem repeats, each composed of 23 amino acids. The most common allele contains 6 repeats, and some alleles may be associated with susceptibility to asthma. Alternatively spliced transcript variants with different 5' UTR, but encoding the same protein, have been found for this gene
Sequence: HQSPKSHFELPHYPGLLAHQKPFIRKSYKCLHKRCRPKLPPSPNNPPKFPNPHQPPKHPDKNSSVVNPTLVATTQIPSVTFPSASTKITTLPNVTFLPQNSpécification
| MUC7 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_152291 | |
| MUC7 | |
| MUC7 (NP_689504,36 a.a. ∽ 135 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 4589 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 7F2 | |
| Mouse monoclonal antibody raised against a partial recombinant MUC7. | |
| MUC7 | |
| DKFZp686J03256/FLJ27047/MG2/MGC34772 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.