missing translation for 'onlineSavingsMsg'
Learn More

MTM1, Mouse, Polyclonal Antibody, Abnova™

Codice prodotto. 16170378
Cambia vista
Click to view available options
Quantity:
50 μg
Dimensione della confezione:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
16170378 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 16170378 Fornitore Abnova N. del fornitore H00004534B01P.50ug

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Mouse polyclonal antibody raised against a full-length human MTM1 protein.

This gene encodes a dual-specificity phosphatase that acts on both phosphotyrosine and phosphoserine. It is required for muscle cell differentiation and mutations in this gene have been identified as being responsible for X-linked myotubular myopathy. [provided by RefSeq

Sequence: MASASTSKYNSHSLENESIKRTSRDGVNRDLTEAVPRLPGETLITDKEVIYICPFNGPIKGRVYITNYRLYLRSLETDSSLILDVPLGVISRIEKMGGATSRGENSYGLDITCKDMRNLRFALKQEGHSRRDMFEILTRYAFPLAHSLPLFAFLNEEKFNVDGWTVYNPVEEYRRQGLPNHHWRITFINKCYELCDTYPALLVVPYRASDDDLRRVATFRSRNRIPVLSWIHPENKTVIVRCSQPLVGMSGKRNKDDEKYLDVIRETNKQISKLTIYDARPSVNAVANKATGGGYESDDAYHNAELFFLDIHNIHVMRESLKKVKDIVYPNVEESHWLSSLESTHWLEHIKLVLTGAIQVADKVSSGKSSVLVHCSDGWDRTAQLTSLAMLMLDSFYRSIEGFEILVQKKWISFGHKFASRIGHGDKNHTDADRSPIFLQFIDCVWQMSKQFPTAFEFNEQFLIIILDHLYSCRFGTFLFNCESARERQKVTERTVSLWSLINSNKEKFKNPFYTKEINRVLYPVASMRHLELWVNYYIRWNPRIKQQQPNPVEQRYMELLALRDEYIKRLEELQLANSAKLSDPPTSPSSPSQMMPHVQTHF

Specifica

Antigen MTM1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human MTM1 protein.
Formulation PBS with no preservative; pH 7.4
Gene MTM1
Gene Accession No. BC030779.1
Gene Alias CNM/MTMX/XLMTM
Gene Symbols MTM1
Host Species Mouse
Immunogen MTM1 (AAH30779.1,1 a.a. ∽ 603 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4534
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Vedi altri risultati Mostra meno risultati

For Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.