Learn More
MTA1, Mouse, Clone: 3A3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant MTA1.
Brand: Abnova H00009112-M09A.200uL
Description
This gene encodes a protein that was identified in a screen for genes expressed in metastatic cells, specifically, mammary adenocarcinoma cell lines. Expression of this gene has been correlated with the metastatic potential of at least two types of carcinomas although it is also expressed in many normal tissues. The role it plays in metastasis is unclear. It was initially thought to be the 70kDa component of a nucleosome remodeling deacetylase complex, NuRD, but it is more likely that this component is a different but very similar protein. These two proteins are so closely related, though, that they share the same types of domains. These domains include two DNA binding domains, a dimerization domain, and a domain commonly found in proteins that methylate DNA. The profile and activity of this gene product suggest that it is involved in regulating transcription and that this may be accomplished by chromatin remodeling. [provided by RefSeq
Sequence: MPSRGLANHGQTRHMGPSRNLLLNGKSYPTKVRLIRGGSLPPVKRRRMNWIDAPDDVFYMATEETRKIRKLLSSSETKRAARRPYKPIALRQSQALPPRPSpecifications
| MTA1 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_004689 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 3A3 | |
| Mouse monoclonal antibody raised against a partial recombinant MTA1. | |
| MTA1 | |
| MTA1 | |
| MTA1 (NP_004680,601 a.a. ∽ 700 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 9112 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.