Learn More
MSX1, Mouse, Clone: 5D11, Abnova™
Mouse monoclonal antibody raised against partial recombinant MSX1.
Brand: Abnova MAB10816.100uL
Description
This gene encodes a member of the muscle segment homeobox gene family. The encoded protein functions as a transcriptional repressor during embryogenesis through interactions with components of the core transcription complex and other homeoproteins. It may also have roles in limb-pattern formation, craniofacial development, particularly odontogenesis, and tumor growth inhibition. Mutations in this gene, which was once known as homeobox 7, have been associated with nonsyndromic cleft lip with or without cleft palate 5, Witkop syndrome, Wolf-Hirschom syndrome, and autosomoal dominant hypodontia. [provided by RefSeq
Specifications
| MSX1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| MSX1 | |
| MSX1 | |
| Recombinant protein corresponding to human MSX1. | |
| RUO | |
| 4487 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Western Blot | |
| 5D11 | |
| Mouse monoclonal antibody raised against partial recombinant MSX1. | |
| In ascites (0.03% sodium azide) | |
| HOX7/HYD1 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.