Learn More
MSN, Mouse, Clone: 41, Abnova™
Mouse monoclonal antibody raised against full-length recombinant MSN.
Brand: Abnova MAB9966.100ug
Description
Moesin (for membrane-organizing extension spike protein) is a member of the ERM family which includes ezrin and radixin. ERM proteins appear to function as cross-linkers between plasma membranes and actin-based cytoskeletons. Moesin is localized to filopodia and other membranous protrusions that are important for cell-cell recognition and signaling and for cell movement. [provided by RefSeq
Specifications
| MSN | |
| Monoclonal | |
| Unconjugated | |
| Western blot (1:500) The optimal working dilution should be determined by the end user. | |
| MSN | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 4478 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 41 | |
| Mouse monoclonal antibody raised against full length recombinant MSN. | |
| In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300) | |
| MSN | |
| Recombinant protein corresponding to full length human MSN. | |
| 100 μg | |
| Primary | |
| It can expression in HeLa whole cell lysate. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.